Orders LinkContactUsLinkComponent
Pressure Bulbs
Pressure Bulbs
Atomiser bulb with pressure valve, red-brown, size 3, PVC, 65
Katalog # 612-9952
Leverandør:   DEUTSCH NEUMANN
Pressure Bulbs
Katalog # 612-9952
Leverandør:  DEUTSCH NEUMANN
Pressure Bulbs
Pressure Bulbs
Atomiser bulb with pressure valve, red-brown, size 3, PVC, 65
Price
177,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Natural rubber, chloroprene or PVC.
Used for filling automatic burettes, Pellet pattern (ball set with net) or atomisers (bulb).
Specifikationer
  • Kapacitet:
    65
  • Beskrivelse:
    Atomiser bulb with pressure valve, red-brown, size 3, PVC
  • Seq:
    1