Lithiumbromid, anhydrous 99+% for analysis
Plastic bottle, 500 g
Leverandør: Thermo Fisher Scientific
Lithiumbromid, anhydrous 99+% for analysis
Katalog # ACRO199875000
Leverandør: Thermo Fisher Scientific
CAS nummer: 7550-35-8
Gå til:
Product Details & Documents
- CAS nummer:7550-35-8
- Formel:LiBr
- MDL nummer:MFCD00011077
- Molecular Weight:86.84 g/mol
- Merck Index:13,05548
- Kogende Pt:1265 °C (760 mmHg)
- Smeltende Pt:550 °C
- Tæthed:3,46 g/cm³
- Opbevaringstemperatur:Stue temperatur
- EINECS:231-439-8
Specifikationer
Specifikationstestresultater
- Appearance:White Powder
- Titration Argentometric:≥99.0 %
- Loss on drying:≤0.5 % (400°C)
- pH:6 to 12 (4 % at 20°C)
- Chloride (Cl):≤1500 ppm
- Carbonate (CO3):≤300 ppm
- Sulfate (SO4):≤200 ppm
- Calcium (Ca):≤50 ppm
- Copper (Cu):≤3 ppm
- Iron (Fe):≤10 ppm
- Magnesium (Mg):≤20 ppm
- Potassium (K):≤300 ppm
- Sodium (Na):≤400 ppm
- Insoluble matter:≤0.005 % (in water)
Specifikationer
- Pakning:Plastic bottle
- Pakn:500 g



