Brom 99.6% for analysis
Glass bottle, plastic-coated, 25 mL
Leverandør: Thermo Fisher Scientific
Brom 99.6% for analysis
Katalog # ACRO196660250
Leverandør: Thermo Fisher Scientific
CAS nummer: 7726-95-6
Gå til:
Product Details & Documents
- CAS nummer:7726-95-6
- Formel:Br₂
- MDL nummer:MFCD00010896
- Molecular Weight:159.81 g/mol
- Merck Index:13,01378
- Kogende Pt:58,7 °C (760 mmHg)
- Smeltende Pt:–7,2 °C
- Tæthed:3,111 g/cm³
- Opbevaringstemperatur:Stue temperatur
- EINECS:231-778-1
- FN:1744
- ADR:8,I
Documents
Certifikater
Søg efter partinummerACRO19666
SDS
Thermo Scientific Stockroom Essentials
Produktlitteratur
Specifikationer
Specifikationstestresultater
- Appearance:Dark red-brown Liquid
- Titration Iodometric:≥99.5 %
- Residue after evaporation:≤100 ppm
- Chlorine (Cl):≤500 ppm
- Iodine (I):≤10 ppm
- Heavy metals (as Pb):≤2 ppm
- Sulfur compounds (as SO4):≤30 ppm
Specifikationer
- Pakning:Glass bottle, plastic-coated
- Pakn:25 mL
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



