Brom 99.6% for analysis
Glass bottle, plastic-coated, 1 L
Leverandør: Thermo Fisher Scientific
Brom 99.6% for analysis
Katalog # ACRO196660010
Leverandør: Thermo Fisher Scientific
CAS nummer: 7726-95-6
Gå til:
Product Details & Documents
- CAS nummer:7726-95-6
- Formel:Br₂
- MDL nummer:MFCD00010896
- Molecular Weight:159.81 g/mol
- Merck Index:13,01378
- Kogende Pt:58,7 °C (760 mmHg)
- Smeltende Pt:–7,2 °C
- Tæthed:3,111 g/cm³
- Opbevaringstemperatur:Stue temperatur
- EINECS:231-778-1
- FN:1744
- ADR:8,I
Documents
Certifikater
Søg efter partinummerACRO19666
SDS
Thermo Scientific Stockroom Essentials
Produktlitteratur
Specifikationer
Specifikationstestresultater
- Appearance:Dark red-brown Liquid
- Titration Iodometric:≥99.5 %
- Residue after evaporation:≤100 ppm
- Chlorine (Cl):≤500 ppm
- Iodine (I):≤10 ppm
- Heavy metals (as Pb):≤2 ppm
- Sulfur compounds (as SO4):≤30 ppm
Specifikationer
- Pakning:Glass bottle, plastic-coated
- Pakn:1 L



