Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Individually Wrapped Serological Pipettes, Falcon®

Individually Wrapped Serological Pipettes, Falcon®

1 ml, 0,01 ml, +, 100 pr. æske
Katalog # 734-0340
Leverandør:   Corning
Individually Wrapped Serological Pipettes, Falcon®
Katalog # 734-0340
Leverandør:  Corning
Individually Wrapped Serological Pipettes, Falcon®
Individually Wrapped Serological Pipettes, Falcon®

1 ml, 0,01 ml, +, 100 pr. æske

Price
kr1,970.00
Log ind for tilgængelighed
1 * 1.000 Items
Product Details & Documents

The pipette graduation alignment system is a fast and simple way to bring graduations to your line of sight. Polystyrene, disposable pipettes for tissue culture, bacteriological, and research applications.

  • Sterile
  • RNase-/DNase-free
  • Noncytotoxic, nonpyrogenicity tested to less than 0,1 EU/ml
  • Color-coded package and markings for ease of identification

Simply pop open or peel back the top of the pipette package and line up the pipettor before completely removing the pipette from its wrapping. When you remove the pipette from its package, the graduations should face you every time.

Packaging: Individually wrapped in plastic-peelable bags, recyclable.

Specifikationer
  • Pakket:
    100 pr. æske
  • Steril:
    +
  • Kapacitet:
    1 ml
  • Seq:
    02
  • Inddeling:
    0,01 ml