2-Propanol 99.5% for analysis
Glass bottle, 1 L
Leverandør: Thermo Fisher Scientific
2-Propanol 99.5% for analysis
Katalog # 184130010.
Leverandør: Thermo Fisher Scientific
CAS nummer: 67-63-0
Gå til:
Product Details & Documents
Synonymer: Isopropylalkohol
- CAS nummer:67-63-0
- Formel:(CH₃)₂CHOH
- MDL nummer:MFCD00011674
- Molecular Weight:60.1 g/mol
- Kogende Pt:81…83 °C (760 mmHg)
- Smeltende Pt:–89,5 °C
- Tæthed:0,785 g/cm³
- Flash Pt:12 °C
- Opbevaringstemperatur:Stue temperatur
- FN:1219
- ADR:3,II
Documents
Specifikationer
Specifikationstestresultater
- Appearance:Clear liquid
- Water:≤0.1 % (Coulometric)
- GC:≥99.5 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.001 %
- Acidity/alkalinity:≤0.0001 meq/g corresp. 0.0007% C3H6O2
- Substance darkened by H2SO4:≤10 APHA
- Cadmium (Cd):≤0.05 ppm
- Calcium (Ca):≤0.5 ppm
- Copper (Cu):≤0.05 ppm
- Iron (Fe):≤0.1 ppm
- Magnesium (Mg):≤0.1 ppm
- Sodium (Na):≤1 ppm
- Sulfur (S):≤0.5 ppm
- Zinc (Zn):≤0.1 ppm
Specifikationer
- Pakning:Glass bottle
- Pakn:1 L
- Cat. No.:184130010.
- Miljøcertificering:Chem21 Solvent Selection Guide recommended after discussion
- Environmentally Preferable:Sikrere sundhed for mennesker og miljø
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



