2-(4-Hydroxyphenyl)ethanol 98%
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
2-(4-Hydroxyphenyl)ethanol 98%
Catalog # ACRO182500250
Supplier: Thermo Fisher Scientific
CAS Number: 501-94-0
Jump to:
Product Details & Documents
Synonyms: NSC 59876, p-HPEA, Tyrosol, 4-Hydroxyphenethyl alcohol
- CAS Number:501-94-0
- Formula:C₈H₁₀O₂
- MDL Number:MFCD00002902
- Molecular Weight:138.17 g/mol
- Boiling Pt:310 °C (1013 hPa)
- Melting Pt:89…92 °C
- EINECS:207-930-8
Specifications
Specification Test Results
- Appearance:Off-white to light beige Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:89 to 93 °C
- GC:≥97.5 %
- HPLC:≥90 %
- Identification:Chlorides: passes test
- UV Lambda max:274 nm
- Absorbance scale:0.75 to 0.83 (A274/A351)
Specifications
- Pakning:Glass bottle for solids
- Pakn:25 g
Specifications
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



