Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
2-(4-Hydroxyphenyl)ethanol 98%

2-(4-Hydroxyphenyl)ethanol 98%

Glass bottle for solids, 5 g
Catalog # ACRO182500050
Irritant
2-(4-Hydroxyphenyl)ethanol 98%
Catalog # ACRO182500050
CAS Number:  501-94-0
2-(4-Hydroxyphenyl)ethanol 98%
2-(4-Hydroxyphenyl)ethanol 98%

Glass bottle for solids, 5 g

CAS Number:  501-94-0
Price
613,00 kr
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  NSC 59876, p-HPEA, Tyrosol, 4-Hydroxyphenethyl alcohol
  • CAS Number:501-94-0
  • Formula:C₈H₁₀O₂
  • MDL Number:MFCD00002902
  • Molecular Weight:138.17 g/mol
  • Boiling Pt:310 °C (1013 hPa)
  • Melting Pt:89…92 °C
  • EINECS:207-930-8

Documents

Specifications

Specification Test Results

  • Appearance:
    Off-white to light beige Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    89 to 93 °C
  • GC:
    ≥97.5 %
  • HPLC:
    ≥90 %
  • Identification:
    Chlorides: passes test
  • UV Lambda max:
    274 nm
  • Absorbance scale:
    0.75 to 0.83 (A274/A351)

Specifications

  • Pakning:
    Glass bottle for solids
  • Pakn:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.