Cumol 99%
Glass bottle, 1 L
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: Cumen
- CAS nummer:98-82-8
- Formel:C₉H₁₂
- MDL nummer:MFCD00008881
- Molecular Weight:120.19 g/mol
- Merck Index:13,02644
- Kogende Pt:151 °C (1013 hPa)
- Smeltende Pt:–96 °C
- Tæthed:0,862 g/cm³
- Flash Pt:31 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:202-704-5
- FN:1918
- ADR:3,III
Specifikationer
Specifikationstestresultater
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4900 to 1.4930 (20°C, 589 nm)
- GC:≥98.5 %
- HPLC:≥97.5 %
Specifikationer
- Pakning:Glass bottle
- Pakn:1 L



