Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Stearinsyre 97%

Plastic bottle, 1 kg
Katalog # ACRO174490010
Leverandør:   Thermo Fisher Scientific
Stearinsyre 97%
Katalog # ACRO174490010
CAS nummer:  57-11-4
Stearinsyre 97%
Stearinsyre 97%

Plastic bottle, 1 kg

CAS nummer:  57-11-4
Price
414,00 kr
Log ind for tilgængelighed
1 * 1 kg
Product Details & Documents
Synonymer:  Octadecanoic acid, Octadecylic acid
  • CAS nummer:57-11-4
  • Formel:C₁₈H₃₆O₂
  • MDL nummer:MFCD00002752
  • Molecular Weight:284.48 g/mol
  • Merck Index:13,08882
  • Kogende Pt:361 °C (760 mmHg)
  • Smeltende Pt:67…69 °C
  • Tæthed:0,84 g/cm³
  • Flash Pt:196 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-313-4

 

Specifikationer

Specifikationstestresultater

  • Appearance:
    White Granular powder or granules or flakes
  • Infrared spectrum:
    Conforms
  • Melting point:
    67 to 71 °C
  • GC:
    ≥96.0 %

Specifikationer

  • Pakning:
    Plastic bottle
  • Pakn:
    1 kg

Specifikationer