Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
L(+)-Cystein 98+%

L(+)-Cystein 98+%

Plastic bottle, 100 g
Katalog # ACRO173601000
Irriterende
Leverandør:   Thermo Fisher Scientific
L(+)-Cystein 98+%
Katalog # ACRO173601000
CAS nummer:  52-90-4
L(+)-Cystein 98+%
L(+)-Cystein 98+%

Plastic bottle, 100 g

CAS nummer:  52-90-4
Price
956,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents
Synonymer:  H-Cys-OH, L-(+)-Cysteine, (R)-2-Amino-3-mercaptopropanoic acid
  • CAS nummer:52-90-4
  • Formel:HSCH₂CH(NH₂)CO₂H
  • MDL nummer:MFCD00064306
  • Molecular Weight:121.16 g/mol
  • Merck Index:13,02810
  • Kogende Pt:254 °C (1013 hPa)
  • Smeltende Pt:220 °C (dec.)
  • Tæthed:1,523 g/cm³ (20 °C)
  • Opbevaringstemperatur:Køleskab
  • EINECS:200-158-2

Documents

Specifikationer

Specifikationstestresultater

  • Appearance:
    White to almost white Powder or crystals
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤0.5 % (105°C, 3 hrs)
  • Solubility:
    (2.5 % in water) Clear colorless
  • Residue after ignition:
    ≤0.1 %
  • pH:
    4.5 to 5.5 (2.5 % in water)
  • Specific optical rotation:
    +7.5° to +9.5° (20°C, 589 nm) (c=8, 5N HCl)
  • Titration with HClO4:
    98.0 to 102.0 %
  • Chloride (Cl):
    ≤0.05 %
  • Sulfate (SO4):
    ≤0.03 %
  • Ammonium (NH4):
    ≤0.02 %
  • Iron (Fe):
    ≤10 ppm
  • Heavy metals (as Pb):
    ≤10 ppm

Specifikationer

  • Pakning:
    Plastic bottle
  • Pakn:
    100 g

Specifikationer