Thionylchlorid 99.5+%
Glass bottle for solids, 500 g
Leverandør: Thermo Fisher Scientific
Thionylchlorid 99.5+%
Katalog # ACRO169495000
Leverandør: Thermo Fisher Scientific
CAS nummer: 7719-09-7
Gå til:
Product Details & Documents
Synonymer: Thionyl dichloride
- CAS nummer:7719-09-7
- Formel:Cl₂OS
- MDL nummer:MFCD00011449
- Molecular Weight:118.97 g/mol
- Merck Index:12,09485
- Kogende Pt:76 °C (1013 hPa)
- Smeltende Pt:–105 °C
- Tæthed:1,64 g/cm³
- Opbevaringstemperatur:Stue temperatur
- EINECS:231-748-8
- FN:1836
- ADR:8,I
Documents
Specifikationer
Specifikationstestresultater
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- GC:≥99.50 %
- HPLC:≥96.0 %
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:500 g
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



