Cyclobutanol 99+%
Glass bottle, 250 mg
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
- CAS nummer:2919-23-5
- Formel:C₄H₈O
- MDL nummer:MFCD00001318
- Molecular Weight:72.11 g/mol
- Kogende Pt:123…124 °C (733 mmHg)
- Tæthed:0,921 g/cm³
- Flash Pt:21 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:220-858-1
- FN:1987
- ADR:3,II
Documents
ACRO16875
SDS
Specifikationer
Specifikationstestresultater
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4340 to 1.4370 (20°C, 589 nm)
- GC:≥99.0 %
Specifikationer
- Pakning:Glass bottle
- Pakn:250 mg
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



