Cyclobutanol 99+%
Glass bottle for solids, 1 g
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
- CAS nummer:2919-23-5
- Formel:C₄H₈O
- MDL nummer:MFCD00001318
- Molecular Weight:72.11 g/mol
- Kogende Pt:123…124 °C (733 mmHg)
- Tæthed:0,921 g/cm³
- Flash Pt:21 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:220-858-1
- FN:1987
- ADR:3,II
Documents
ACRO16875
SDS
Specifikationer
Specifikationstestresultater
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4340 to 1.4370 (20°C, 589 nm)
- GC:≥99.0 %
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:1 g



