Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Cyclohexan 99+% for spectroscopy

Cyclohexan 99+% for spectroscopy

Glass bottle, 500 mL
Katalog # ACRO167745000
Irriterende
Sundhed
Miljø
Brandfarlig
Leverandør:   Thermo Fisher Scientific
Cyclohexan 99+% for spectroscopy
Katalog # ACRO167745000
CAS nummer:  110-82-7
Cyclohexan 99+% for spectroscopy
Cyclohexan 99+% for spectroscopy

Glass bottle, 500 mL

CAS nummer:  110-82-7
Price
585,00 kr
Log ind for tilgængelighed
1 * 500 mL
Product Details & Documents
Synonymer:  Hexahydrobenzene, Hexamethylene
  • CAS nummer:110-82-7
  • Formel:C₆H₁₂
  • MDL nummer:MFCD00003814
  • Molecular Weight:84.16 g/mol
  • Merck Index:13,02752
  • Kogende Pt:81 °C (1013 hPa)
  • Smeltende Pt:6,5 °C
  • Tæthed:0,77 g/cm³
  • Flash Pt:–18 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:203-806-2
  • FN:1145
  • ADR:3,II

Documents

Specifikationer

Specifikationstestresultater

  • Appearance:
    Clear liquid
  • Water:
    ≤0.01 %
  • GC:
    ≥99.0 %
  • UV:
    at 210 nm A: ≤1
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.0005 %
  • Acidity:
    ≤0.0002 meq/g

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    500 mL

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.