Orders LinkContactUsLinkComponent
Magnetic stirring bars, plain, PTFE
Magnetic stirring bars, plain, PTFE
15 mm, 6 mm
Katalog # COWI001.215.6
Leverandør:   COWIE
Magnetic stirring bars, plain, PTFE
Katalog # COWI001.215.6
Leverandør:  COWIE
Magnetic stirring bars, plain, PTFE
Magnetic stirring bars, plain, PTFE
15 mm, 6 mm
Price
15,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Plain PTFE magnetic stirring bars have similar performance to cylindrical bars, but provide more turbulence at low speeds.
Specifikationer
  • Ø:
    6 mm
  • Længde:
    15 mm
  • Seq:
    02