Pipette tips, universal, Corning®
Affaset åbning, rund, 1 - 200 µl, -, Transparent, Bulk
Leverandør: Corning
Pipette tips, universal, Corning®
Katalog # 613-4997
Leverandør: Corning
Gå til:
Product Details & Documents
Corning® universal fit tips are designed to provide a reliable fit with all major brand pipettors.
- Beveled orifice helps ensure accurate fluid delivery
- All tips are certified RNase-/DNase-free and non pyrogenic
- Stack rack tips feature a stack of five racks, each containing 96 tips, for a total of 480 tips in a space-saving design
- Bulk packed tips are non sterile and very economical
1 to 200 µl universal fit tips are graduated at the 10 µl, 50 µl, and 100 µl volumes.
Documents
Specifikationer
- Pakket:Bulk
- Steril:-
- Volumen:1 - 200 µl
- Seq:05
- Spidstype:Affaset åbning, rund
- Spits farge:Transparent
- Cat. No.:613-4997
- Miljøcertificering:I-RECs, ISO 14001 - Reynosa
- Environmentally Preferable:Lav fremstillingspåvirkning