Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
DL-Alanin 99%

DL-Alanin 99%

Plastic bottle, 100 g
Katalog # ACRO159091000
Leverandør:   Thermo Fisher Scientific
DL-Alanin 99%
Katalog # ACRO159091000
CAS nummer:  302-72-7
DL-Alanin 99%
DL-Alanin 99%

Plastic bottle, 100 g

CAS nummer:  302-72-7
Price
420,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents
Synonymer:  (±)-Alanine, DL-α-Alanine, H-DL-Ala-OH, α-Alanine
  • CAS nummer:302-72-7
  • Formel:C₃H₇NO₂
  • MDL nummer:MFCD00064408
  • Molecular Weight:89.09 g/mol
  • Merck Index:13,00203
  • Kogende Pt:189 °C (1013 hPa)
  • Smeltende Pt:272…275 °C (dec.)
  • Tæthed:1,42 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:206-126-4

Documents

Specifikationer

Specifikationstestresultater

  • Appearance:
    White Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤0.2 % (105°C, 3 hrs)
  • Sulfated ash:
    ≤0.1 %
  • UV transmittance:
    ≥95 % (C=10, H2O, 430 nm) 1 cm cell
  • Titration with HClO4:
    98.5 to 101.5 %
  • Chloride (Cl):
    ≤200 ppm
  • Sulfate (SO4):
    ≤300 ppm
  • Ammonium (NH4):
    ≤200 ppm
  • Iron (Fe):
    ≤20 ppm
  • Arsenic oxide (As2O3):
    ≤1 ppm
  • Heavy metals (as Pb):
    ≤10 ppm
  • Other amino acids:
    ≤0.5 % (TLC)

Specifikationer

  • Pakning:
    Plastic bottle
  • Pakn:
    100 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.