Orders LinkContactUsLinkComponent
Centrifugerør, Corning®
50 ml, PP, Skruelåg, 15500×g, koniske, 25 stk./rack
Katalog # 734-1868
   Lav fremstillingspåvirkning
Leverandør:   Corning
Centrifugerør, Corning®
Katalog # 734-1868
Leverandør:  Corning
   Lav fremstillingspåvirkning
Centrifugerør, Corning®
Centrifugerør, Corning®
50 ml, PP, Skruelåg, 15500×g, koniske, 25 stk./rack
Price
1.980,00 kr
Log ind for tilgængelighed
1 * 500 Items
Product Details & Documents
Corning 50 ml centrifuge tubes made of PP or PET.
  • Conical, with HDPE CentriStar™ or plug seal caps or without cap, sterile
  • Black printed graduations and a large white marking spot
  • Certified non pyrogenic and DNase/RNase-free
The CentriStar™ cap with leakproof design has a easy-on/easy-off flat top and offers advanced ergonomics with its wider knurls and roll-over edge design for easier gripping.
 : Skumracks og CentriStar™-låg fås separat.
 : Bulk or in racks.
Specifikationer
  • Lukkestørrelse:
    Skruelåg
  • Pakket:
    25 stk./rack
  • Model:
    koniske
  • Kapacitet:
    50 ml
  • Materiale:
    PP
  • Seq:
    04
  • RCF maks.:
    15500×g
  • Cat. No.:
    734-1868
  • Miljøcertificering:
    I-RECs, ISO 14001 - Reynosa
  • Environmentally Preferable:
    Lav fremstillingspåvirkning