Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Centrifugerør, Corning®

Centrifugerør, Corning®

50 ml, PET, Skruelåg, 3600×g, koniske, 25 stk./rack
Katalog # 734-1829
   Lav fremstillingspåvirkning
Leverandør:   Corning
Centrifugerør, Corning®
Katalog # 734-1829
Leverandør:  Corning
   Lav fremstillingspåvirkning
Centrifugerør, Corning®
Centrifugerør, Corning®

50 ml, PET, Skruelåg, 3600×g, koniske, 25 stk./rack

Price
kr2,860.00
Log ind for tilgængelighed
1 * 500 Items
Product Details & Documents

Corning 50 ml centrifuge tubes made of PP or PET.

  • Conical, with HDPE CentriStar™ or plug seal caps or without cap, sterile
  • Black printed graduations and a large white marking spot
  • Certified non pyrogenic and DNase/RNase-free

The CentriStar™ cap with leakproof design has a easy-on/easy-off flat top and offers advanced ergonomics with its wider knurls and roll-over edge design for easier gripping.

Bestillingsoplysninger: Skumracks og CentriStar™-låg fås separat.

Emballering: Bulk or in racks.

Specifikationer
  • Lukkestørrelse:
    Skruelåg
  • Pakket:
    25 stk./rack
  • Model:
    koniske
  • Kapacitet:
    50 ml
  • Materiale:
    PET
  • Seq:
    03
  • RCF maks.:
    3600×g
  • Cat. No.:
    734-1829
  • Miljøcertificering:
    I-RECs, ISO 14001 - Reynosa
  • Environmentally Preferable:
    Lav fremstillingspåvirkning