Triethylamin 99%, pure
Glass bottle, 1 L
Leverandør: Thermo Fisher Scientific
Triethylamin 99%, pure
Katalog # ACRO157910010
Leverandør: Thermo Fisher Scientific
CAS nummer: 121-44-8
Gå til:
Product Details & Documents
Synonymer: (Diethylamino)ethane, TEA
- CAS nummer:121-44-8
- Formel:(C₂H₅)₃N
- MDL nummer:MFCD00009051
- Molecular Weight:101.19 g/mol
- Merck Index:13,09740
- Kogende Pt:90 °C (760 mmHg)
- Smeltende Pt:–115 °C
- Tæthed:0,728 g/cm³
- Flash Pt:–11 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:204-469-4
- FN:1296
- ADR:3,II
Documents
Certifikater
Søg efter partinummerACRO15791
SDS
Thermo Scientific Stockroom Essentials
Product Literature
Specifikationer
Specifikationstestresultater
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.3990 to 1.4010 (20°C, 589 nm)
- Water:≤0.2 % (K.F.)
- GC:≥98.5 %
- Color scale:≤15 APHA
Specifikationer
- Pakning:Glass bottle
- Pakn:1 L
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



