96-well plates, polypropylene, without lids
Opbevaringsplade, 96-brønds, klar, rund brønd, 360 µl, -, 25/pakke, 4 pakker/æske
Leverandør: Corning
Gå til:
Product Details & Documents
Corning polypropylene microplates offer both small volume and large volume (blocks) well formats to meet assay and storage requirements.
- Flat, round, or V-shaped well bottom
- Features uniform skirt heights for greater robotic gripping surface
- Solvent resistant polypropylene provides compatibility with many common organic solvents (e.g., DMSO, ethanol, methanol)
- RNase-/DNase-free
- Available sterile or non sterile
Bestillingsoplysninger: Plates are packaged 25 per pack, 4 packs per case. Nonsterile 450 µL plate is packaged 10 plates per bag and 5 bags per case.
Specifikationer
- Pakket:25/pakke, 4 pakker/æske
- Steril:-
- Beskrivelse:Opbevaringsplade, 96-brønds, klar, rund brønd
- Seq:06
- Brøndvolumen:360 µl