Orders LinkContactUsLinkComponent
96-well plates, polypropylene, without lids
96-well plates, polypropylene, without lids
Opbevaringsplade, 96-brønds, klar, V-formet brønd, 320 µl, +, 25/pakke, 4 pakker/æske
Katalog # 734-1545
Leverandør:   Corning
96-well plates, polypropylene, without lids
Katalog # 734-1545
Leverandør:  Corning
96-well plates, polypropylene, without lids
96-well plates, polypropylene, without lids
Opbevaringsplade, 96-brønds, klar, V-formet brønd, 320 µl, +, 25/pakke, 4 pakker/æske
Price
3.380,00 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents
Corning polypropylene microplates offer both small volume and large volume (blocks) well formats to meet assay and storage requirements.
  • Flat, round, or V-shaped well bottom
  • Features uniform skirt heights for greater robotic gripping surface
  • Solvent resistant polypropylene provides compatibility with many common organic solvents (e.g., DMSO, ethanol, methanol)
  • RNase-/DNase-free
  • Available sterile or non sterile
Bestillingsoplysninger: Plates are packaged 25 per pack, 4 packs per case. Nonsterile 450 µL plate is packaged 10 plates per bag and 5 bags per case.
Specifikationer
  • Pakket:
    25/pakke, 4 pakker/æske
  • Steril:
    +
  • Beskrivelse:
    Opbevaringsplade, 96-brønds, klar, V-formet brønd
  • Seq:
    01
  • Brøndvolumen:
    320 µl