Thermometer test caps
TX24L, test cap, Lumberg type, +3 °C
Katalog # 620-0378
Leverandør: COMARK
Gå til:
Product Details & Documents
Certified test capsules can be used for quick checking of the thermometer's function against specifications. They are connected to the thermometer instead of a sensor and consist of a resistor that corresponds exactly to a given temperature.
Specifikationer
- Område:+3 °C
- Beskrivelse:TX24L, test cap, Lumberg type