Thermometer probes
Type T high temperature air probe up to +400 °C, sub-miniature connector, AK33M, −100…+400 °C, - mm
Catalog # 620-0334
Supplier: COMARK
Jump to:
Product Details & Documents
Specifications
- Område:−100…+400 °C
- Beskrivelse:Type T high temperature air probe up to +400 °C, sub-miniature connector, AK33M
- Ø×L:- mm