Thermometer probes
Flexible wire type K air probe, with sub-miniature connector, AK29M, −100…+250 °C, - mm
Katalog # COMAAK29M
Leverandør: COMARK
Gå til:
Product Details & Documents
Specifikationer
- Område:−100…+250 °C
- Beskrivelse:Flexible wire type K air probe, with sub-miniature connector, AK29M
- Ø×L:- mm