Anisol 99%, pure
Glass bottle, 250 mL
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: Methyl phenyl ether, Methoxybenzene
- CAS nummer:100-66-3
- Formel:C₇H₈O
- MDL nummer:MFCD00008354
- Molecular Weight:108.14 g/mol
- Merck Index:13,00672
- Kogende Pt:154 °C (760 mmHg)
- Smeltende Pt:–37 °C
- Tæthed:0,99 g/cm³
- Flash Pt:45 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:202-876-1
- FN:2222
- ADR:3,III
Documents
Specifikationer
Specifikationstestresultater
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5160 to 1.5190 (20°C, 589 nm)
- Water:≤0.1 % (K.F.)
- GC:≥98.5 %
Specifikationer
- Pakning:Glass bottle
- Pakn:250 mL



