Anisol 99%, pure
Glass bottle, 5 mL
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: Methyl phenyl ether, Methoxybenzene
- CAS nummer:100-66-3
- Formel:C₇H₈O
- MDL nummer:MFCD00008354
- Molecular Weight:108.14 g/mol
- Merck Index:13,00672
- Kogende Pt:154 °C (760 mmHg)
- Smeltende Pt:–37 °C
- Tæthed:0,99 g/cm³
- Flash Pt:45 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:202-876-1
- FN:2222
- ADR:3,III
Documents
Specifikationer
Specifikationstestresultater
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5160 to 1.5190 (20°C, 589 nm)
- Water:≤0.1 % (K.F.)
- GC:≥98.5 %
Specifikationer
- Pakning:Glass bottle
- Pakn:5 mL
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



