Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Anisol 99%, pure

Anisol 99%, pure

Glass bottle, 1 L
Catalog # ACRO153920010
Flammable
Irritant
Anisol 99%, pure
Catalog # ACRO153920010
CAS Number:  100-66-3
Anisol 99%, pure
Anisol 99%, pure

Glass bottle, 1 L

CAS Number:  100-66-3
Price
637,00 kr
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Methyl phenyl ether, Methoxybenzene
  • CAS Number:100-66-3
  • Formula:C₇H₈O
  • MDL Number:MFCD00008354
  • Molecular Weight:108.14 g/mol
  • Merck Index:13,00672
  • Boiling Pt:154 °C (760 mmHg)
  • Melting Pt:–37 °C
  • Density:0,99 g/cm³
  • Flash Pt:45 °C
  • Storage Temperature:Stue temperatur
  • EINECS:202-876-1
  • UN:2222
  • ADR:3,III

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5160 to 1.5190 (20°C, 589 nm)
  • Water:
    ≤0.1 % (K.F.)
  • GC:
    ≥98.5 %

Specifications

  • Pakning:
    Glass bottle
  • Pakn:
    1 L

Specifications