Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Formaldehyde sodium bisulphite 95%

Formaldehyde sodium bisulphite 95%

Plastic bottle, 1 kg
Katalog # ACRO150690010
Leverandør:   Thermo Fisher Scientific
Formaldehyde sodium bisulphite 95%
Katalog # ACRO150690010
CAS nummer:  870-72-4
Formaldehyde sodium bisulphite 95%
Formaldehyde sodium bisulphite 95%

Plastic bottle, 1 kg

CAS nummer:  870-72-4
Price
712,00 kr
Log ind for tilgængelighed
1 * 1 kg
Product Details & Documents

Appearance: White to off-white Powder

Synonymer:  Sodium Hydroxymethanesulphonate, Hydroxymethanesulphonic acid sodium salt
  • CAS nummer:870-72-4
  • Formel:CH₄O₄S
  • MDL nummer:MFCD00044664
  • Molecular Weight:134.08 g/mol
  • Smeltende Pt:200 °C
  • Flash Pt:184 °C
  • Opbevaringstemperatur:Stue temperatur

Documents

Specifikationer

Specifikationstestresultater

  • Appearance:
    White to off-white Powder
  • Infrared spectrum:
    Conforms
  • Titration:
    ≥94 %
  • HPLC:
    ≥97.5 %

Specifikationer

  • Pakning:
    Plastic bottle
  • Pakn:
    1 kg

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.