(S)-(-)-Æblesyre 99%
Plastic bottle, 100 g
Leverandør: Thermo Fisher Scientific
(S)-(-)-Æblesyre 99%
Katalog # ACRO150591000
Leverandør: Thermo Fisher Scientific
CAS nummer: 97-67-6
Gå til:
Product Details & Documents
Synonymer: L(-)-Malic acid, (S)-(-)-Hydroxy-1,4-butanedioic acid, (S)-(-)-Hydroxysuccinic acid, (S)-2-hydroxysuccinic acid, L(-)-Hydroxysuccinic acid, L(-)-Hydroxy-1,4-butanedioic acid, L-(–)-Malic Acid
- CAS nummer:97-67-6
- Formel:C₄H₆O₅
- MDL nummer:MFCD00064213
- Molecular Weight:134.09 g/mol
- Merck Index:13,05730
- Kogende Pt:322 °C (1013 hPa)
- Smeltende Pt:99…107 °C
- Tæthed:1,595 g/cm³ (20 °C)
- Flash Pt:220 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:202-601-5
Specifikationer
Specifikationstestresultater
- Appearance:White Crystalline powder or granules
- Infrared spectrum:Conforms
- Melting point:99 to 107 °C
- Sulfated ash:≤0.2 %
- Specific optical rotation:-1.5° to -3° (20 °C, 589 nm) (c=8.5, water)
- Titration with NaOH:≥98.5 %
- Chloride (Cl):≤100 ppm
- Sulfate (SO4):≤300 ppm
- Arsenic (As):≤2 ppm
- Heavy metals (as Pb):≤20 ppm
- Fumaric acid:≤0.2 %
Specifikationer
- Pakning:Plastic bottle
- Pakn:100 g



