2-Propanol 99.5+%
Metal drum, 25 L
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: Isopropylalkohol
- CAS nummer:67-63-0
- Formel:(CH₃)₂CHOH
- MDL nummer:MFCD00011674
- Molecular Weight:60.1 g/mol
- Merck Index:13,05228
- Kogende Pt:82 °C (1013 hPa)
- Smeltende Pt:–89,5 °C
- Tæthed:0,785 g/cm³
- Flash Pt:12 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-661-7
- FN:1219
- ADR:3,II
- NÅ:01-2119457558-25
Specifikationer
Specifikationstestresultater
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Water:≤0.2 % (K.F.)
- GC:≥99.50 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.005 %
Specifikationer
- Pakning:Metal drum
- Pakn:25 L



