Pentylacetat 99%
Glass bottle, 250 mL
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: n-Pentyl acetate, Acetic acid amyl ester, Acetic acid pentyl ester, Pentyl acetate
- CAS nummer:628-63-7
- Formel:C₇H₁₄O₂
- MDL nummer:MFCD00009500
- Molecular Weight:130.19 g/mol
- Kogende Pt:149 °C (760 mmHg)
- Smeltende Pt:–70,8 °C
- Tæthed:0,87 g/cm³
- Flash Pt:24 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:211-047-3
- FN:1104
- ADR:3,III
Documents
Specifikationer
Specifikationstestresultater
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4010 to 1.4030 (20°C, 589 nm)
- GC:≥98.5 %
- TLC:≥97.5 %
- Specific optical rotation:-10.8° to -11.8° (20°C, 589 nm) (c=2 in DMF)
Specifikationer
- Pakning:Glass bottle
- Pakn:250 mL



