2-Hexanon 98%
Glass bottle, 500 mL
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: Butylmethylketon
- CAS nummer:591-78-6
- Formel:C₆H₁₂O
- MDL nummer:MFCD00009482
- Molecular Weight:100.16 g/mol
- Merck Index:13,06060
- Kogende Pt:127 °C (760 mmHg)
- Smeltende Pt:–57 °C
- Tæthed:0,81 g/cm³
- Flash Pt:23 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:209-731-1
- FN:1224
- ADR:3,III
Documents
ACRO14688
SDS
Specifikationer
Specifikationstestresultater
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3995 to 1.4015 (20°C, 589 nm)
- GC:≥97.5 %
Specifikationer
- Pakning:Glass bottle
- Pakn:500 mL
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



