2-Hexanon 98%
Glass bottle, 100 mL
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: Butylmethylketon
- CAS nummer:591-78-6
- Formel:C₆H₁₂O
- MDL nummer:MFCD00009482
- Molecular Weight:100.16 g/mol
- Merck Index:13,06060
- Kogende Pt:127 °C (760 mmHg)
- Smeltende Pt:–57 °C
- Tæthed:0,81 g/cm³
- Flash Pt:23 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:209-731-1
- FN:1224
- ADR:3,III
Specifikationer
Specifikationstestresultater
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3995 to 1.4015 (20°C, 589 nm)
- GC:≥97.5 %
Specifikationer
- Pakning:Glass bottle
- Pakn:100 mL