L(+)-Valin 99%
Plastic bottle, 500 g
: H-Val-OH, Valine, (S)-(+)-Valine
- :72-18-4
- :C₅H₁₁NO₂
- :MFCD00064220
- :117.15 g/mol
- :13,09458
- :217 °C (1013 hPa)
- :315 °C
- :1,23 g/cm³
- :Stue temperatur
- :200-773-6
Documents
ACRO14081
SDS
- Appearance:White Powder or crystals
- Infrared spectrum:Conforms
- Loss on drying:≤0.3 %
- Sulfated ash:≤0.1 %
- Specific optical rotation:+26.5° to +29.0° (20°C, 589 nm) (c=8, 6 N HCl)
- Titration with HClO4:≥98.5 %
- Chloride (Cl):≤500 ppm
- Sulfate (SO4):≤300 ppm
- Ammonium (NH4):≤200 ppm
- Iron (Fe):≤30 ppm
- Heavy metals (as Pb):≤10 ppm
- Pakning:Plastic bottle
- Pakn:500 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



