L(+)-Valin 99%
Plastic bottle, 100 g
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: H-Val-OH, Valine, (S)-(+)-Valine
- CAS nummer:72-18-4
- Formel:C₅H₁₁NO₂
- MDL nummer:MFCD00064220
- Molecular Weight:117.15 g/mol
- Merck Index:13,09458
- Kogende Pt:217 °C (1013 hPa)
- Smeltende Pt:315 °C
- Tæthed:1,23 g/cm³
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-773-6
Documents
ACRO14081
SDS
Specifikationer
Specifikationstestresultater
- Appearance:White Powder or crystals
- Infrared spectrum:Conforms
- Loss on drying:≤0.3 %
- Sulfated ash:≤0.1 %
- Specific optical rotation:+26.5° to +29.0° (20°C, 589 nm) (c=8, 6 N HCl)
- Titration with HClO4:≥98.5 %
- Chloride (Cl):≤500 ppm
- Sulfate (SO4):≤300 ppm
- Ammonium (NH4):≤200 ppm
- Iron (Fe):≤30 ppm
- Heavy metals (as Pb):≤10 ppm
Specifikationer
- Pakning:Plastic bottle
- Pakn:100 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



