Tyramin 97%
Glass bottle for solids, 5 g
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: 4-Hydroxyphenethylamine, 4-(2-Aminoethyl)phenol
- CAS nummer:51-67-2
- Formel:C₈H₁₁NO
- MDL nummer:MFCD00008193
- Molecular Weight:137.18 g/mol
- Kogende Pt:175…181 °C (8 mmHg)
- Smeltende Pt:158…163 °C
- EINECS:200-115-8
- FN:3263
Documents
ACRO14061
SDS
Specifikationer
Specifikationstestresultater
- Appearance:Off-white to beige to light brown Powder
- Infrared spectrum:Conforms
- Melting point:158 to 163 °C
- Titration with HClO4:≥96.0 %
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:5 g



