L(-)-Tryptophan 99%
Plastic bottle, 100 g
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: L-Tryptophan, Tryptophan, L-α-Amino-3-indolepropionic acid, (S)-2-Amino-3-(3-indolyl)propionic acid, H-Trp-OH
- CAS nummer:73-22-3
- Formel:C₁₁H₁₂N₂O₂
- MDL nummer:MFCD00064340
- Molecular Weight:204.23 g/mol
- Merck Index:12,09929
- Smeltende Pt:280…285 °C (dec.)
- Tæthed:1,34 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-795-6
Documents
Specifikationer
Specifikationstestresultater
- Appearance:White to yellowish-white Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.5 % (1 g, 105°C)
- Specific optical rotation:-29.0° to -33.0° (20°C, 589 nm) (C=1, H2O)
- Titration with HClO4:98.5 to 101.5 % (On dry substance)
Specifikationer
- Pakning:Plastic bottle
- Pakn:100 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



