L(-)-Tryptophan 99%
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: L-Tryptophan, Tryptophan, L-α-Amino-3-indolepropionic acid, (S)-2-Amino-3-(3-indolyl)propionic acid, H-Trp-OH
- CAS Number:73-22-3
- Formula:C₁₁H₁₂N₂O₂
- MDL Number:MFCD00064340
- Molecular Weight:204.23 g/mol
- Merck Index:12,09929
- Melting Pt:280…285 °C (dec.)
- Density:1,34 g/cm³ (20 °C)
- Storage Temperature:Stue temperatur
- EINECS:200-795-6
Documents
Specifications
Specification Test Results
- Appearance:White to yellowish-white Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.5 % (1 g, 105°C)
- Specific optical rotation:-29.0° to -33.0° (20°C, 589 nm) (C=1, H2O)
- Titration with HClO4:98.5 to 101.5 % (On dry substance)
Specifications
- Pakning:Glass bottle for solids
- Pakn:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



