1,2,4-Triazol 99.5%
Plastic bottle, 25 g
Leverandør: Thermo Fisher Scientific
1,2,4-Triazol 99.5%
Katalog # ACRO139280250
Leverandør: Thermo Fisher Scientific
CAS nummer: 288-88-0
Gå til:
Product Details & Documents
Synonymer: 1,2,4-1H-Triazole, 1H-1,2,4-Triazole
- CAS nummer:288-88-0
- Formel:C₂H₃N₃
- MDL nummer:MFCD00005228
- Molecular Weight:69.07 g/mol
- Merck Index:12,09735
- Kogende Pt:260 °C (760 mmHg)
- Smeltende Pt:119…122 °C
- Tæthed:1,394 g/cm³ (25 °C)
- Flash Pt:140 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:206-022-9
Documents
ACRO13928
SDS
Specifikationer
Specifikationstestresultater
- Appearance:White Powder and/or flakes
- Infrared spectrum:Conforms
- Melting point:119 to 122 °C
- Water:≤0.2 % (K.F.)
- HPLC:≥99.4 %
Specifikationer
- Pakning:Plastic bottle
- Pakn:25 g



