Spermidin 99%
Glass bottle for solids, 5 g
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: N1-(3-Aminopropyl)butane-1,4-diamine, N-(3-Aminopropyl)-1,4-diaminobutane, 4-Azaoctamethylenediamine
- CAS nummer:124-20-9
- Formel:C₇H₁₉N₃
- MDL nummer:MFCD00008229
- Molecular Weight:145.25 g/mol
- Merck Index:12,08893
- Kogende Pt:128…130 °C (14 mmHg)
- Smeltende Pt:22…25 °C
- Tæthed:0,925 g/cm³
- Flash Pt:>110 °C
- Opbevaringstemperatur:Køleskab
- EINECS:204-689-0
- FN:3259
- ADR:8,II
Documents
Specifikationer
Specifikationstestresultater
- Appearance:Clear colorless Liquid after melting
- Infrared spectrum:Conforms
- Refractive index:1.4785 to 1.4805 (20°C, 589 nm)
- GC:≥98.5 %
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:5 g



