Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Propyl 4-hydroxybenzoat 99+%

Propyl 4-hydroxybenzoat 99+%

Plastic bottle, 500 g
Katalog # ACRO131595000
Irriterende
Leverandør:   Thermo Fisher Scientific
Propyl 4-hydroxybenzoat 99+%
Katalog # ACRO131595000
CAS nummer:  94-13-3
Propyl 4-hydroxybenzoat 99+%
Propyl 4-hydroxybenzoat 99+%

Plastic bottle, 500 g

CAS nummer:  94-13-3
Price
965,00 kr
Log ind for tilgængelighed
1 * 500 g
Product Details & Documents
Synonymer:  Propyl 4-hydroxybenzoate, Propyl parahydroxybenzoate, Propylparaben, 4-Hydroxybenzoic acid propyl ester
  • CAS nummer:94-13-3
  • Formel:C₁₀H₁₂O₃
  • MDL nummer:MFCD00002354
  • Molecular Weight:180.2 g/mol
  • Merck Index:13,07958
  • Kogende Pt:292 °C (1013 hPa)
  • Smeltende Pt:95…99 °C
  • Tæthed:1,28 g/cm³ (20 °C)
  • Flash Pt:180 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:202-307-7
Specifikationer

Specifikationstestresultater

  • Appearance:
    White to almost white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    95 to 99 °C
  • Sulfated ash:
    ≤0.1 %
  • GC:
    ≥99.0 %
  • Appearance of solution:
    (10% in ethanol 96%) clear colorless to

Specifikationer

  • Pakning:
    Plastic bottle
  • Pakn:
    500 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.