Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Propyl 4-hydroxybenzoat 99+%

Propyl 4-hydroxybenzoat 99+%

Glass bottle for solids, 5 g
Katalog # ACRO131590050
Irriterende
Leverandør:   Thermo Fisher Scientific
Propyl 4-hydroxybenzoat 99+%
Katalog # ACRO131590050
CAS nummer:  94-13-3
Propyl 4-hydroxybenzoat 99+%
Propyl 4-hydroxybenzoat 99+%

Glass bottle for solids, 5 g

CAS nummer:  94-13-3
Price
kr311.00
Log ind for tilgængelighed
1 * 5 g
Product Details & Documents
Synonymer:  Propyl 4-hydroxybenzoate, Propyl parahydroxybenzoate, Propylparaben, 4-Hydroxybenzoic acid propyl ester
  • CAS nummer:94-13-3
  • Formel:C₁₀H₁₂O₃
  • MDL nummer:MFCD00002354
  • Molecular Weight:180.2 g/mol
  • Merck Index:13,07958
  • Kogende Pt:292 °C (1013 hPa)
  • Smeltende Pt:95…99 °C
  • Tæthed:1,28 g/cm³ (20 °C)
  • Flash Pt:180 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:202-307-7

Documents

Specifikationer

Specifikationstestresultater

  • Appearance:
    White to almost white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    95 to 99 °C
  • Sulfated ash:
    ≤0.1 %
  • GC:
    ≥99.0 %
  • Appearance of solution:
    (10% in ethanol 96%) clear colorless to

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    5 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.