Stearinsyre, Millipore®
Plastic bottle for solids, 100 g
Katalog # 569398-100
Leverandør: Merck Millipore (Calbiochem)
Stearinsyre, Millipore®
Katalog # 569398-100
Leverandør: Merck Millipore (Calbiochem)
CAS nummer: 57-11-4
Gå til:
Product Details & Documents
Synonymer: Octadecanoic acid, Octadecylic acid
- CAS nummer:57-11-4
- Formel:C₁₈H₃₆O₂
- MDL nummer:MFCD00002752
- Molecular Weight:284.48 g/mol
- Merck Index:13,08882
- Kogende Pt:361 °C (1013 hPa)
- Smeltende Pt:67 °C
- Tæthed:0,94 g/cm³ (20 °C)
- Flash Pt:196 °C (closed cup)
- Opbevaringstemperatur:Køleskab
- EINECS:200-313-4
Specifikationer
Specifikationer
- Pakning:Plastic bottle for solids
- Pakn:100 g



