1,3-Propansulton 97%
Glass bottle, 25 g
Supplier: Thermo Fisher Scientific
1,3-Propansulton 97%
Catalog # ACRO131420250
Supplier: Thermo Fisher Scientific
CAS Number: 1120-71-4
Jump to:
Product Details & Documents
Synonyms: 1,2-Oxathiolane 2,2-dioxide
- CAS Number:1120-71-4
- Formula:C₃H₆O₃S
- MDL Number:MFCD00005355
- Molecular Weight:122.14 g/mol
- Boiling Pt:180 °C (30 mmHg)
- Melting Pt:30…33 °C
- Density:1,392 g/cm³
- Flash Pt:>110 °C
- Storage Temperature:Stue temperatur
- EINECS:214-317-9
- UN:2811
- ADR:6.1,III
Documents
ACRO13142
SDS
Specifications
Specification Test Results
- Appearance:White to light yellow Low melting solid or liquid
- Infrared spectrum:Conforms
- Melting point:28 to 33 °C
- GC:≥96.0 %
Specifications
- Pakning:Glass bottle
- Pakn:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



