Phthalsyre 99%
Plastic bottle, 1 kg
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: Pathalic acid
- CAS nummer:88-99-3
- Formel:C₈H₆O₄
- MDL nummer:MFCD00002467
- Molecular Weight:166.13 g/mol
- Merck Index:12,07527
- Kogende Pt:315 - 317 °C (1013 hPa)
- Smeltende Pt:200…205 °C (dec.)
- Tæthed:1,576 g/cm³ (20 °C)
- Flash Pt:168 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:201-873-2
Specifikationer
Specifikationstestresultater
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:200 to 205 °C (Dec)
- Titration with NaOH:≥98.5 %
- Chloride (Cl):≤50 ppm
- Sulfate (SO4):≤50 ppm
- Iron (Fe):≤50 ppm
- Heavy metals (as Pb):≤10 ppm
Specifikationer
- Pakning:Plastic bottle
- Pakn:1 kg



