1-Naphtol 99+%
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: α-Naphtol
- CAS Number:90-15-3
- Formula:C₁₀H₈O
- MDL Number:MFCD00003930
- Molecular Weight:144.17 g/mol
- Merck Index:13,06409
- Boiling Pt:278…280 °C (760 mmHg)
- Melting Pt:95…97 °C
- Density:1,224 g/cm³ (20 °C)
- Flash Pt:125 °C
- Storage Temperature:Stue temperatur
- EINECS:201-969-4
- UN:2811
- ADR:6.1,III
Documents
Specifications
Specification Test Results
- Appearance:Beige to grey to brown Crystalline powder or crystals or flakes
- Infrared spectrum:Conforms
- Melting point:95 to 97 °C
- GC:≥99.0 %
Specifications
- Pakning:Glass bottle for solids
- Pakn:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



