Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Melatonin 99%

Glass bottle, 1 g
Katalog # ACRO125360010
Leverandør:   Thermo Fisher Scientific
Melatonin 99%
Katalog # ACRO125360010
CAS nummer:  73-31-4
Melatonin 99%
Melatonin 99%

Glass bottle, 1 g

CAS nummer:  73-31-4
Price
947,00 kr
Log ind for tilgængelighed
1 * 1 g
Product Details & Documents
Synonymer:  Melatonin, N-(2-(5-Methoxy-1H-indol-3-yl)ethyl)acetamide
  • CAS nummer:73-31-4
  • Formel:C₁₃H₁₆N₂O₂
  • MDL nummer:MFCD00005655
  • Molecular Weight:232.28 g/mol
  • Kogende Pt:512,8 °C
  • Smeltende Pt:118 °C
  • Flash Pt:264 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-797-7
Specifikationer

Specifikationstestresultater

  • Appearance:
    White to cream to yellow Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    116 to 120 °C
  • Loss on drying:
    ≤0.5 % (1 g, 105 °C)
  • Heavy metals:
    ≤20 ppm
  • HPLC:
    ≥98.5 %

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    1 g

Specifikationer