Diethylether 99+% stabilised, pure
Metal drum, 5 L
Supplier: Thermo Fisher Scientific
Diethylether 99+% stabilised, pure
Catalog # ACRO123990050
Supplier: Thermo Fisher Scientific
CAS Number: 60-29-7
Jump to:
Product Details & Documents
Synonyms: Ether
- CAS Number:60-29-7
- Formula:(CH₃CH₂)₂O
- MDL Number:MFCD00011646
- Molecular Weight:74.12 g/mol
- Merck Index:12,03852
- Boiling Pt:34,6 °C (760 mmHg)
- Melting Pt:–116 °C
- Density:0,714 g/cm³
- Flash Pt:–45 °C
- UN:1155
- ADR:3,I
- :stabiliseret med BHT/Ionol
- Storage Temperature:Stue temperatur
- EINECS:200-467-2
- REACH:01-2119535785-29
Documents
Certificates
Search by Lot NumberACRO12399
SDS
ACRO123990010_TDS_EN ACRO123990025_TDS_EN ACRO123990050_TDS_EN
Product specification
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.3510 to 1.3540 (20°C, 589 nm)
- Water:≤0.2 % (Coulometric)
- GC:≥99.0 %
- Color scale:≤10 APHA
- Stabilizer:5 to 10 ppm BHT
- Residue after evaporation:≤0.002 %
- Peroxides:≤0.001 %
Specifications
- Pakning:Metal drum
- Pakn:5 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



